accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1435-43-4

FFCl

CAS RN 1435-43-4

IUPAC Name: 1-chloro-3,5-difluorobenzene
Synonyms: 1-chloro-3,5-difluorobenzene
Molecular Weight (g/mol): 148.54
Molecular Formula: C6H3ClF2
InChi Key: RFKBODCWHNDUTJ-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results