accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14324-55-1

ZnH3CNH3CSSH3CNCH3SS

CAS RN 14324-55-1

IUPAC Name: zinc(2+) bis((diethylcarbamothioyl)sulfanide)
Synonyms: zinc(2+) didiethyldithiocarbamate
Molecular Weight (g/mol): 361.9
Molecular Formula: C10H20N2S4Zn
InChi Key: RKQOSDAEEGPRER-UHFFFAOYSA-L
SMILES: [Zn++].CCN(CC)C([S-])=S.CCN(CC)C([S-])=S
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)