accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OO(R)H3CNH2HCl

CAS RN 14316-06-4

IUPAC Name: hydrogen methyl 2-aminopropanoate chloride
Synonyms: hydrogen methyl 2-aminopropanoate chloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(C)N
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)