accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SFFFFFOHOCl

CAS RN 1431329-70-2

IUPAC Name: 2-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 2-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 282.61
Molecular Formula: C7H4ClF5O2S
InChi Key: KNKMPGVWOPIPMH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results