accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14293-78-8

OOOKV

CAS RN 14293-78-8

IUPAC Name: potassium vanadium trioxidandiide
Synonyms: potassium vanadium trioxidandiide
Molecular Weight (g/mol): 138.04
Molecular Formula: KO3V
InChi Key: WAMGQABPPBZHJS-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[K+].[V]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results