accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14284-93-6

OOOOOOH3CCH3CH3H3CH3CH3CRu

CAS RN 14284-93-6

IUPAC Name: tris(pentane-2,4-dione) ruthenium
Synonyms: ruthenium(III) acetylacetonate
Molecular Weight (g/mol): 401.42
Molecular Formula: C15H24O6Ru
InChi Key: GNHBILLJFGEMKL-UHFFFAOYSA-N
SMILES: [Ru].CC(=O)CC(C)=O.CC(=O)CC(C)=O.CC(=O)CC(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results