CAS RN 142738-94-1
IUPAC Name:
1-methoxy-3-(trifluoromethoxy)benzene
Synonyms:
1-methoxy-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
192.14
Molecular Formula:
C8H7F3O2
InChi Key:
OKSRAPDSZKICKZ-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
3-(Trifluoromethoxy)anisole, 97+%, Thermo Scientific™
CAS: 142738-94-1 Molecular Formula: C8H7F3O2 Molecular Weight (g/mol): 192.137 MDL Number: MFCD01631504 InChI Key: OKSRAPDSZKICKZ-UHFFFAOYSA-N Synonym: 3-trifluoromethoxy anisole,1-methoxy-3-trifluoromethoxy benzene,benzene, 1-methoxy-3-trifluoromethoxy,trifluoro 3-methoxyphenoxy methane,pubchem4158,3-trifluoromethoxyanisole,acmc-1c79q,ksc494o1j,1-methoxy-3-trifluoromethoxy-benzene,1-methoxy-3-trifluoromethyloxy benzene PubChem CID: 2777188 IUPAC Name: 1-methoxy-3-(trifluoromethoxy)benzene SMILES: COC1=CC(=CC=C1)OC(F)(F)F