accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1426315-84-5

NOFF

CAS RN 1426315-84-5

IUPAC Name: N-benzyl-2,6-difluoro-N-phenylbenzamide
Synonyms: N-benzyl-2,6-difluoro-N-phenylbenzamide
Molecular Weight (g/mol): 323.34
Molecular Formula: C20H15F2NO
InChi Key: GPQFSRTYCAFMCY-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results