accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14243-65-3

AuPBrPhPhPh

CAS RN 14243-65-3

IUPAC Name: gold(1+) triphenylphosphane bromide
Synonyms: gold(1+) triphenylphosphine bromide
Molecular Weight (g/mol): 539.16
Molecular Formula: C18H15AuBrP
InChi Key: UIYUJCNGYSSFGP-UHFFFAOYSA-M
SMILES: [Br-].[Au+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results