accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14242-01-4

OPOOOEr

CAS RN 14242-01-4

IUPAC Name: erbium(3+) hydrate phosphate
Synonyms: erbium(3+) hydrate phosphate
Molecular Weight (g/mol): 280.24
Molecular Formula: ErH2O5P
InChi Key: PDQXJPLNEUYGOX-UHFFFAOYSA-K
SMILES: O.[Er+3].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results