accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3NH2OH3C

CAS RN 14227-17-9

IUPAC Name: 2,4,6-trimethoxyaniline
Synonyms: 2,4,6-trimethoxyaniline
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: FNSAKXLEFPFZOM-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(N)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results