accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NNNNN

CAS RN 14213-12-8

IUPAC Name: 3-(1H-1,2,3,4-tetrazol-1-yl)aniline
Synonyms: 3-(1,2,3,4-tetrazol-1-yl)aniline
Molecular Weight (g/mol): 161.17
Molecular Formula: C7H7N5
InChi Key: YXWXLBHIUVBLIH-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)N1C=NN=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results