accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNNOH3CBr(?)

CAS RN 1421261-61-1

IUPAC Name: 3-(4-bromophenyl)-4-(2-methoxyphenyl)-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-(2-methoxyphenyl)-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 344.21
Molecular Formula: C16H14BrN3O
InChi Key: CCXAVTXHPGUSRI-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1N1C(C)=NN=C1C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results