accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COCH3O

CAS RN 142-92-7

IUPAC Name: hexyl acetate
Synonyms: hexyl acetate N-hexyl acetate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: AOGQPLXWSUTHQB-UHFFFAOYSA-N
SMILES: CCCCCCOC(C)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)