CAS RN 14177-46-9
CAS RN 14177-46-9
IUPAC Name:
manganese(2+) dioxotungstenbis(olate)
Synonyms:
manganese(2+) tungstate
Molecular Weight (g/mol):
302.77
Molecular Formula:
MnO4W
InChi Key:
CRLHSBRULQUYOK-UHFFFAOYSA-N
SMILES:
[Mn++].[O-][W]([O-])(=O)=O
1
–
1
of
1
results
1
Manganese(II) tungsten oxide, 99.9% (metals basis)
CAS: 14177-46-9 MDL Number: MFCD00014212 Synonym: Manganese tungstate