accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrP

CAS RN 14114-05-7

IUPAC Name: cyclopropyltriphenylphosphanium bromide
Synonyms: cyclopropyltriphenylphosphanium bromide
Molecular Weight (g/mol): 383.27
Molecular Formula: C21H20BrP
InChi Key: XMPWFKHMCNRJCL-UHFFFAOYSA-M
SMILES: [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)