accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 141091-38-5

BOOH3CCH3H3CH3CCH3

CAS RN 141091-38-5

IUPAC Name: 2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-decyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 268.25
Molecular Formula: C16H33BO2
InChi Key: HRMCLNRSAPIJDP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCB1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results