accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14092-04-7

CH3MgBr(E)

CAS RN 14092-04-7

IUPAC Name: magnesium(2+) prop-1-en-1-ide bromide
Synonyms: magnesium(2+) prop-1-en-1-ide bromide
Molecular Weight (g/mol): 145.28
Molecular Formula: C3H5BrMg
InChi Key: WQMNNHNWITXOAH-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC=[CH-]
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results