accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14077-58-8

H3COClO

CAS RN 14077-58-8

IUPAC Name: 2-ethoxyacetyl chloride
Synonyms: ethoxy-acetyl chloride
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
SMILES: CCOCC(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results