accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFBrFFF

CAS RN 140690-56-8

IUPAC Name: 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 307.03
Molecular Formula: C9H5BrF6
InChi Key: SWFFFUJOWAJJCH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CBr)C(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results