accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14024-64-7

CH3OH3COHCH3OH3COHOTi(E/Z)(E/Z)

CAS RN 14024-64-7

IUPAC Name: bis(4-hydroxypent-3-en-2-one); oxotitanium
Synonyms: bis(4-hydroxypent-3-en-2-one); titanium(II) oxide
Molecular Weight (g/mol): 264.1
Molecular Formula: C10H16O5Ti
InChi Key: ADVORQMAWLEPOI-UHFFFAOYSA-N
SMILES: O=[Ti].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results