accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14024-58-9

OOOOMnH3CH3CCH3CH3

CAS RN 14024-58-9

IUPAC Name: bis(4-hydroxypent-3-en-2-one) dihydrate manganese
Synonyms: bis(4-hydroxypent-3-en-2-one) dihydrate manganese
Molecular Weight (g/mol): 291.2
Molecular Formula: C10H20MnO6
InChi Key: XBBNRHYLXGCQDT-UHFFFAOYSA-N
SMILES: O.O.[Mn].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results