accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 140187-25-3

IUPAC Name: 2-amino-3-(5-iodo-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)propanoic acid
Synonyms: iodo-willardiine
Molecular Weight (g/mol): 325.06
Molecular Formula: C7H8IN3O4
InChi Key: AXXYLTBQIQBTES-UHFFFAOYNA-N
SMILES: NC(CN1C=C(I)C(=O)NC1=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results