accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CeOClOOOOClOOOOClOOO

CAS RN 14017-47-1

IUPAC Name: cerium(3+) triperchlorate
Synonyms: cerium(3+) triperchlorate
Molecular Weight (g/mol): 438.45
Molecular Formula: CeCl3O12
InChi Key: LJBTWTBUIINKRU-UHFFFAOYSA-K
SMILES: [Ce+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results