accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OSOO

CAS RN 139926-23-1

IUPAC Name: 3-ethoxythiophene-2-carboxylate
Synonyms: 3-ethoxythiophene-2-carboxylate
Molecular Weight (g/mol): 171.19
Molecular Formula: C7H7O3S
InChi Key: FTKDCOINUAYGTC-UHFFFAOYSA-M
SMILES: CCOC1=C(SC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results