accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13991-37-2

CH3OHO(E)

CAS RN 13991-37-2

IUPAC Name: (2E)-pent-2-enoic acid
Synonyms: trans-2-pentenoic acid E-2-pentencarbonsaeure pent-2t-ensaeure trans-pent-2-ensaeure trans-α,β-penteneoic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: YIYBQIKDCADOSF-ONEGZZNKSA-N
SMILES: CC\C=C\C(O)=O
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)