accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13988-67-5

H3CH3CCH3OO

CAS RN 13988-67-5

IUPAC Name: 4,4-dimethyl-1-phenylpentane-1,3-dione
Synonyms: 4,4-dimethyl-1-phenylpentane-1,3-dione
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: HORVLKADAZQYRS-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results