accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3C(R)(R)OOH3CCH3(S)CH3OO(S)NHOHNOHO(R)H3COO

CAS RN 1397-94-0

IUPAC Name: (2R,3S,6S,7R,8R)-3-(3-formamido-2-hydroxybenzamido)-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl 3-methylbutanoate
Synonyms: antimycin A antipiricullin antimycin A1 fintrol virosin antimycin A1b
Molecular Weight (g/mol): 548.63
Molecular Formula: C28H40N2O9
InChi Key: UIFFUZWRFRDZJC-SBOOETFBSA-N
SMILES: CCCCCC[C@@H]1[C@@H](OC(=O)CC(C)C)[C@H](C)OC(=O)[C@@H](NC(=O)C2=CC=CC(NC=O)=C2O)[C@@H](C)OC1=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results