accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CBrNH2

CAS RN 139305-96-7

IUPAC Name: 1-(3-bromophenyl)ethan-1-amine
Synonyms: 1-(3-bromophenyl)ethanamine
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: LIBZHYLTOAGURM-UHFFFAOYNA-N
SMILES: CC(N)C1=CC(Br)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results