accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13922-41-3

IUPAC Name: (naphthalen-1-yl)boronic acid
Synonyms: naphthalen-1-ylboronic acid
Molecular Weight (g/mol): 171.99
Molecular Formula: C10H9BO2
InChi Key: HUMMCEUVDBVXTQ-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=CC=C1
Quantity 
  • (2)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results