accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1391008-15-3

IUPAC Name: 2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 267.18
Molecular Formula: C10H9F4NO3
InChi Key: POFOCSWADMUTGH-UHFFFAOYNA-N
SMILES: NC(CC1=C(F)C=C(OC(F)(F)F)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)