accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 139092-78-7

NNNN

CAS RN 139092-78-7

IUPAC Name: 4-(9H-carbazol-9-yl)-N,N-bis[4-(9H-carbazol-9-yl)phenyl]aniline
Synonyms: 4-(carbazol-9-yl)-N,N-bis[4-(carbazol-9-yl)phenyl]aniline
Molecular Weight (g/mol): 740.91
Molecular Formula: C54H36N4
InChi Key: AWXGSYPUMWKTBR-UHFFFAOYSA-N
SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results