accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13896-65-6

RuIII

CAS RN 13896-65-6

IUPAC Name: ruthenium(3+) triiodide
Synonyms: ruthenium(3+) triiodide
Molecular Weight (g/mol): 481.78
Molecular Formula: I3Ru
InChi Key: LJZVDOUZSMHXJH-UHFFFAOYSA-K
SMILES: [Ru+3].[I-].[I-].[I-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results