accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONH(S)OHO

CAS RN 138662-63-2

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-diphenylpropanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3,3-diphenylpropanoic acid
Molecular Weight (g/mol): 341.41
Molecular Formula: C20H23NO4
InChi Key: TYJDOLCFYZSNQC-KRWDZBQOSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results