accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13862-78-7

NH3NH3SHSHSWS

CAS RN 13862-78-7

IUPAC Name: disulfanylidenetungsten diamine disulfanide
Synonyms: tungsten disulfide diamine disulfanide
Molecular Weight (g/mol): 348.16
Molecular Formula: H8N2S4W
InChi Key: CCFCMIWYHMKPAO-UHFFFAOYSA-L
SMILES: N.N.[SH-].[SH-].S=[W]=S
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)