accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH3NH3NH3NH3NH3ClClClCo

CAS RN 13859-51-3

IUPAC Name: cobalt(3+) pentaamine trichloride
Synonyms: chloropentamminecobalt chloride
Molecular Weight (g/mol): 250.44
Molecular Formula: Cl3CoH15N5
InChi Key: NSYALVBBDKTCLE-UHFFFAOYSA-K
SMILES: N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)