CAS RN 138485-81-1
CAS RN 138485-81-1
IUPAC Name:
3-[4-(methylsulfanyl)phenyl]propanoic acid
Synonyms:
3-[4-(methylsulfanyl)phenyl]propanoic acid
Molecular Weight (g/mol):
196.26
Molecular Formula:
C10H12O2S
InChi Key:
ASTVFRXZLBGVRL-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(CCC(O)=O)C=C1
1
–
1
of
1
results
1
3-[4-(Methylthio)phenyl]propionic acid, 96%
CAS: 138485-81-1 Molecular Formula: C10H12O2S Molecular Weight (g/mol): 196.264 MDL Number: MFCD03425696 InChI Key: ASTVFRXZLBGVRL-UHFFFAOYSA-N PubChem CID: 3801098 IUPAC Name: 3-(4-methylsulfanylphenyl)propanoic acid SMILES: CSC1=CC=C(C=C1)CCC(=O)O