accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13844-89-8

KKNNNNOOOOOOOOPd

CAS RN 13844-89-8

IUPAC Name: dipotassium palladium tetranitrite
Synonyms: dipotassium palladium tetranitrite
Molecular Weight (g/mol): 368.64
Molecular Formula: K2N4O8Pd
InChi Key: QZDIEKDUJYMSIK-UHFFFAOYSA-J
SMILES: [K+].[K+].[Pd].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results