accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13823-50-2

KNH2SOOO

CAS RN 13823-50-2

IUPAC Name: potassium sulfamate
Synonyms: potassium sulfamate
Molecular Weight (g/mol): 135.18
Molecular Formula: H2KNO3S
InChi Key: BTAAXEFROUUDIL-UHFFFAOYSA-M
SMILES: [K+].NS([O-])(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)