CAS RN 138182-21-5
IUPAC Name:
(2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
(2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
329.73
Molecular Formula:
C14H16ClNO6
InChi Key:
OQWBAXBVBGNSPW-MBJXGIAVSA-N
SMILES:
OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O
1
–
1
of
1
results
1
6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%
CAS: 138182-21-5 Molecular Formula: C14H16ClNO6 Molecular Weight (g/mol): 329.73 MDL Number: MFCD00467206,MFCD01310928 InChI Key: OQWBAXBVBGNSPW-MBJXGIAVSA-N Synonym: salmon-gal,rose-gal,6-chloro-3-indolyl beta-d-galactopyranoside PubChem CID: 44630036 IUPAC Name: (2S,3S,4S,5R,6S)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O