accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13817-37-3

H2OH2OH2OH2OFFCo

CAS RN 13817-37-3

IUPAC Name: λ²-cobalt(2+) tetrahydrate difluoride
Synonyms: λ²-cobalt(2+) tetrahydrate difluoride
Molecular Weight (g/mol): 168.99
Molecular Formula: CoF2H8O4
InChi Key: MNHUTFNEBYCZAF-UHFFFAOYSA-L
SMILES: O.O.O.O.[F-].[F-].[Co++]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results