accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1378524-41-4

CH3FNNH3COOCH3OCH3OCH3(E/Z)

CAS RN 1378524-41-4

IUPAC Name: N-[3-(2-fluorophenyl)-2-methylprop-2-en-1-yl]-3,4,5-trimethoxy-N-[2-(1-methylpyrrolidin-2-yl)ethyl]benzamide
Synonyms: N-[3-(2-fluorophenyl)-2-methylprop-2-en-1-yl]-3,4,5-trimethoxy-N-[2-(1-methylpyrrolidin-2-yl)ethyl]benzamide
Molecular Weight (g/mol): 470.59
Molecular Formula: C27H35FN2O4
InChi Key: JVRRRKAXHMGUHZ-UHFFFAOYNA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(=O)N(CCC1CCCN1C)CC(C)=CC1=CC=CC=C1F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results