accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13770-89-3

IUPAC Name: nickel(2+) disulfamate
Synonyms: nickel bis(sulfamidate)
Molecular Weight (g/mol): 250.85
Molecular Formula: H4N2NiO6S2
InChi Key: KERTUBUCQCSNJU-UHFFFAOYSA-L
SMILES: [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
Physical Form 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)