accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFEr

CAS RN 13760-83-3

IUPAC Name: erbium(3+) trifluoride
Synonyms: erbium(3+) trifluoride
Molecular Weight (g/mol): 224.25
Molecular Formula: ErF3
InChi Key: QGJSAGBHFTXOTM-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Er+3]
Grade 
  • (4)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)