CAS RN 1374659-33-2
CAS RN 1374659-33-2
IUPAC Name:
3,5-dichloro-4-(difluoromethyl)pyridine
Synonyms:
3,5-dichloro-4-(difluoromethyl)pyridine
Molecular Weight (g/mol):
197.99
Molecular Formula:
C6H3Cl2F2N
InChi Key:
SQQNYQZGLSIAIG-UHFFFAOYSA-N
SMILES:
FC(F)C1=C(Cl)C=NC=C1Cl
1
–
1
of
1
results
1
3,5-Dichloro-4-(difluoromethyl)pyridine, 97%, Thermo Scientific™
CAS: 1374659-33-2 Molecular Formula: C6H3Cl2F2N Molecular Weight (g/mol): 197.994 MDL Number: MFCD22124922 InChI Key: SQQNYQZGLSIAIG-UHFFFAOYSA-N Synonym: 3,5-dichloro-4-difluoromethyl pyridine PubChem CID: 60623001 IUPAC Name: 3,5-dichloro-4-(difluoromethyl)pyridine SMILES: C1=C(C(=C(C=N1)Cl)C(F)F)Cl