accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1374516-07-0

CH3FOOOH

CAS RN 1374516-07-0

IUPAC Name: 3-[4-({4-fluoro-4'-methyl-[1,1'-biphenyl]-2-yl}methoxy)phenyl]propanoic acid
Synonyms: 3-[4-({4-fluoro-4'-methyl-[1,1'-biphenyl]-2-yl}methoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 364.42
Molecular Formula: C23H21FO3
InChi Key: LPGBXHWIQNZEJB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=C(COC2=CC=C(CCC(O)=O)C=C2)C=C(F)C=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results