accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1373921-05-1

CH3OBrFFFF

CAS RN 1373921-05-1

IUPAC Name: 1-(bromomethyl)-2-fluoro-5-methoxy-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-fluoro-5-methoxy-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 287.05
Molecular Formula: C9H7BrF4O
InChi Key: RISYNKJCXPMUQS-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(CBr)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results