accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFFFNH2

CAS RN 1373920-87-6

IUPAC Name: 1-[2-methyl-5-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[2-methyl-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C9H10F3NO
InChi Key: HUHITEUBBNSNER-UHFFFAOYSA-N
SMILES: CC1=C(CN)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results