accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1373920-86-5

H3COFFCl

CAS RN 1373920-86-5

IUPAC Name: 1-(6-chloro-2,3-difluorophenyl)ethan-1-one
Synonyms: 1-(6-chloro-2,3-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: MSWPVZQPVHBWRL-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results