accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1373920-76-3

CH3OOH2NFFFF

CAS RN 1373920-76-3

IUPAC Name: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzamide
Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 237.15
Molecular Formula: C9H7F4NO2
InChi Key: YAJGTXLKIJZBFG-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(=C1)C(N)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results